EMAIL THIS PAGE TO A FRIEND
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology
Selective Ablation of GIRK Channels in Dopamine Neurons Alters Behavioral Effects of Cocaine in Mice.
PMID 27468917
Abstract
The increase in dopamine (DA) neurotransmission stimulated by in vivo cocaine exposure is tempered by G protein-dependent inhibitory feedback mechanisms in DA neurons of the ventral tegmental area (VTA). G protein-gated inwardly rectifying K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1675
Picrotoxin, powder
C30H34O13