EMAIL THIS PAGE TO A FRIEND
Acta neurologica Scandinavica
N-terminal ACTH fragments increase the CSF beta-EP content in Alzheimer type dementia.
PMID 2845700
Abstract
Eleven patients with presenile Alzheimer type dementia (ATD) were treated with N-terminal ACTH fragments for 14 days. No change in cognitive functions was observed during the treatment. A significant increase in CSF beta-endorphin (beta-EP) levels was found, while ACTH and beta-lipoprotein remain unaffected. The possibility that ACTH and its moieties could interfere with beta-EP activities in CNS is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2407
Adrenocorticotropic Hormone Fragment 1-17 human, rat, ≥97% (HPLC)
C95H145N29O23S