EMAIL THIS PAGE TO A FRIEND
Talanta
Intracellular DNA and microRNA sensing based on metal-organic framework nanosheets with enzyme-free signal amplification.
PMID 28501216
Abstract
A new fluorescent sensing platform based on ultrathin metal-organic framework (MOF) nanosheets (MnDMS) was prepared from the flexible ligand 2,2-dimethylsuccinate and Mn ions. The MnDMS nanoparticles can be obtained by simply ultrasonication of the MnDMS crystal, and then can be exfoliated into nanosheets by Li-intercalation method. The MnDMS nanosheets can be easily assembled with biological probes, leading to efficient fluorescence quenching of the fluorophore tagged ssDNA and microRNA (miRNA). By using a hybridization chain reaction (HCR) strategy, the fluorescence signal can be obviously amplified. A good linearity was obtained from 1 pM to 200 pM of target ssDNA, with a detection limit of 0.2 pM. The HCR/MnDMS system provides an effective way to monitor miRNA in living cells. Therefore, the MnDMS nanosheets can be used as a new kind of platform in biomedical sensing applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D186007
2,2-Dimethylsuccinic acid, 99%
C6H10O4