EMAIL THIS PAGE TO A FRIEND
Plant physiology
Higher Novel L-Cys Degradation Activity Results in Lower Organic-S and Biomass in
PMID 28743765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS020792
O-Acetyl-L-serine
C5H9NO4