EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
Effects of dopamine receptor antagonists on gastrin and vomiting responses to apomorphine.
PMID 2888024
Abstract
Apomorphine (0.05 mg/kg intravenously) was given to conscious dogs, and gastrin levels were measured in peripheral venous blood with a radioimmunoassay. Apomorphine induced an increase of gastrin levels which peaked at 5 min. The peripheral dopamine D-2/DA2 receptor antagonist domperidone (0.2 mg/kg), but not halopemide (0.1-1 mg/kg) nor the D-1/DA1 receptor antagonist SCH 23,390 (0.1 mg/kg), blocked the gastrin response to apomorphine. Both domperidone and halopemide, but not SCH 23,390, blocked the apomorphine-induced vomiting. These results suggest that apomorphine increases gastrin levels by an action at D-2/DA2 receptors, which are situated outside the blood brain barrier and differ from the receptor inducing the vomiting.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3041
Halopemide, ≥98% (HPLC)
C21H22ClFN4O2