EMAIL THIS PAGE TO A FRIEND
Royal Society open science
Improved surface-enhanced Raman and catalytic activities of reduced graphene oxide-osmium hybrid nano thin films.
PMID 28989743
Abstract
Reduced graphene oxide-osmium (rGO-Os) hybrid nano dendtrites have been prepared by simple liquid/liquid interface method for the first time. The method involves the introduction of phase-transfered metal organic precursor in toluene phase and GO dispersion in the aqueous phase along with hydrazine hydrate as the reducing agent. Dendritic networks of Os nanoparticles and their aggregates decorating rGO layers are obtained. The substrate shows improved catalytic and surface-enhanced activities comparable with previous reports. The catalytic activity was tested for the reduction of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
398594
Osmium(III) chloride, 99.9% trace metals basis
Cl3Os