EMAIL THIS PAGE TO A FRIEND
Acta histochemica. Supplementband
Regression of the interdigital tissue during the formation of the digits.
PMID 3085151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201677
Janus Green B, certified by the Biological Stain Commission, Dye content 65 %
C30H31ClN6