MilliporeSigma
Photoreactors
Photoreactors provide an all-in-one platform to consistently and reproducibly conduct photoredox catalyzed reactions. Each photochemical reactor consists of a light source and a cooling system to maintain stability of the reactants and products.
Our photoreactors provide versatile and efficient means for performing photochemical reactions from screening to scale-up, with a range of wavelengths, and options allowing multiple photocatalytic reactions to be run in parallel.
  • SynLED Parallel Photoreactor 2.0 – next generation photoredox parallel synthesis
  • Penn PhD Photoreactor M2 – user-defined parameters including temperature, light intensity, fan speed and stirring
In addition, explore our broad portfolio of photocatalysts, including acridinium, iridium, and ruthenium catalysts, visible light metal complex photoredox catalysts, and metal-free organic catalysts, for all your photoredox chemistry needs.
Sort by Relevance
Showing 1-14 of 14
Change View
Showing 1-14 of 14
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-14 of 14 results
Page 1 of 1
Page 1 of 1
SynLED 2.0 Photoreactor with cover removed and powered on showing the blue light source emitting from the reactor.
SynLED Parallel Photoreactor
The SynLED Parallel Photoreactor 2.0 houses a 4 x 4 reaction block lit from the bottom with either 365 nm or 450 nm wavelength LEDs. The newly designed reactor adds improved heat sink technology and a performance quartz LED cover to protect the light source from solvents while still delivering excellent light penetration. These upgrades allow even higher levels of consistency across reactions and between runs.
A Penn PhD Photoreactor M2 with a white arched top emitting blue light, placed on a lab bench with industrial equipment in the background.
Penn PhD Photoreactor M2
The Penn PhD Photoreactor is a complete photoredox catalysis reactor.  This benchtop instrument includes a variable LED light source (365 to 450 nm), mechanical stirring and cooling.  The reactor supports various vial sizes including gc, 4, 8, 20, and 40 mL.
Related Product Resources
Page 1 of 3
Related Webinars
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Latest Innovations in Photoredox Catalysis
In this webinar you will learn the versatility of photoredox catalysis, recent advancements in the use of commercial photoreactors, strategies for optimizing reaction conditions and extending photoredox catalysis to a broader range of nucleophiles.
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Photoredox Catalysis for Efficient Reactions
In this webinar you will explore how photocatalysis is influencing green chemistry, from the design of advanced catalysts to their potential in industrial applications across sectors such as pharmaceuticals and the synthesis of essential compounds.
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Fundamentals and Applications of Photoredox Catalysis in Organic and Macromolecular Synthesis Chemistry World
Watch the insightful webinar to discover the fundamentals and applications of photoredox catalysts in organic and macromolecular synthesis.