MilliporeSigma
Size Exclusion Chromatography Resins
Two white plastic containers with black caps used for storing liquids.
Size exclusion chromatography (SEC) is widely used in purification of native or recombinant proteins, viruses, and plasma-derived biotherapeutics. This separation method is not restricted by buffer composition. 
Our SEC resin offerings include:
  • Fractogel® EMD BioSEC resin for efficient separation of dimers, high molecular weight aggregates, purification of viruses, and polishing of blood plasma factors.
Products
Sort by Relevance
Showing 1-2 of 2
Change View
Showing 1-2 of 2
You have viewed 1-2 of 2 results
Page 1 of 1
Page 1 of 1