Microcon® Centrifugal Filters Applications Guidelines & Specifications
Microcon® Centrifugal Filters simply and efficiently concentrate and desalt solutions of DNA, RNA, protein or other macromolecules, using any centrifuge that can accept 1.5 mL tubes. With the low- binding Ultracel® membrane, Microcon® filters offer:
- Typical recoveries of >95%, even for dilute solutions
- Reverse spin to maximize recovery, even in the smallest samples
- Convenient storage of filtrate or concentrated sample in standard microfuge tube.
The Microcon® DNA Fast Flow Filter is optimized for the concentration and recovery of genomic DNA. The low non-specific binding characteristics of the membrane and the other device components, coupled with its medical-grade o-ring seal, allows the device to accommodate several wash steps with minimal sample loss. With its inverted recovery spin, the device can concentrate samples up to 100X (for greatest consistency and reproducibility, we recommend concentration factors of < 20X).
Applications Guidelines
Use the following table to choose the correct Microcon® device for your application.
| Microcon® Device | |||
|---|---|---|---|
| Application | 10K | 30K | DNA Fast Flow |
| Peptide and growth factor concentration | ● | ||
| Protein concentration and desalting of columns eluates | ● | ● | |
| Protein concentration before electrophoresis or other assays | ● | ● | |
| Protein removal prior to HPLC | ● | ● | |
| Purification of macromolecular components found in tissue culture extracts and cell lysates | ● | ● | |
| Concentration of biological samples (antigens, antibodies, enzymes) | ● | ||
| Concentration and desalting of nucleic acids (single-or double-stranded) | ● | ● | ● |
| Removal of labeled nucleotides | ● | ● | ● |
| Removal of labeled amino acids | ● | ● | ● |
| Removal of primers from amplified DNA | ● | ● | |
| Removal of linkers prior to cloning | ● | ● | |
Specifications | |
|---|---|
| Maximum initial sample volume | 0.5 mL (500 μL) |
| Typical final concentrate volume | 5–50 μL |
| Maximum relative centrifugal force | |
| Microcon® 10K devices | 14,000 × g |
| Microcon® 30K devices | 14,000 × g |
| Microcon® DNA Fast Flow devices | 500 × g |
| Active membrane area | 0.32 cm2 |
| Hold-up volume | ≤ 10 μL |
Ordering Information
Microcon® Centrifugal Filters
Response not successful: Received status code 500