Fortfahren mit
137279
L-Arginin
EMPROVE® EXPERT, EP, USP
Synonym(e):
(S)-2-Amino-5-guanidinopentansäure
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
Lineare Formel:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS-Nummer:
Molekulargewicht:
174.20
UNSPSC Code:
12352209
NACRES:
NA.26
eCl@ss:
32160406
MDL number:
Beilstein/REAXYS Number:
1725413
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
biological source
non-animal source
Quality Segment
agency
EP, USP
product line
EMPROVE® EXPERT
assay
>98.5%
form
crystalline powder
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, residual solvents
mp
222 °C (dec.) (lit.)
solubility
H2O: 100 mg/mL
suitability
suitable for manufacturing use
application(s)
pharmaceutical (small molecule)
SMILES string
N[C@@H](CCCNC(N)=N)C(O)=O
InChI
1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1
InChI key
ODKSFYDXXFIFQN-BYPYZUCNSA-N
Gene Information
human ... NOS1(4842), NOS2(4843)
rat ... Ppm1a(24666)
General description
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.
M-Clarity Program
As part of our Emprove® Program, our raw materials are offered with Emprove® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.
Application
L-Arginine is a non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-Arginine monohydrochloride is a modified version of L-Arginine with improved solubility.
Biochem/physiol Actions
Substrat von Stickstoffmonoxid-Synthase, das zu Citrullin und Stickstoffmonoxid (NO) umgesetzt wird. Induziert Insulinfreisetzung über einen Stickstoffmonoxid-abhängigen Mechanismus.
Legal Information
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
11009A4474A8094
L-Arginin EMPROVE® EXPERT, EP, USP
SAFC
137279
L-Arginin
L-Arginin
SAFC
A4474
L-Arginin
description
EMPROVE® EXPERT, EP, USP
description
BioUltra, ≥99.5% (NT)
description
-
description
from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
product line
EMPROVE® EXPERT
product line
BioUltra
product line
-
product line
BioXtra
agency
USP, EP
agency
-
agency
-
agency
meets EP testing specifications, USP/NF, meets USP testing specifications
Quality Level
500
Quality Level
100
Quality Level
400
Quality Level
300
form
crystalline powder
form
powder or crystals
form
crystalline powder
form
powder
impurities
endotoxin, residual solvents
impurities
insoluble matter, passes filter test, ≤0.3% foreign amino acids
impurities
endotoxin, heavy metals, trace metals, residual solvents, tested
impurities
endotoxin, tested
solubility
H2O: 100 mg/mL
solubility
H2O: 0.5 M at 20 °C, clear, colorless
solubility
H2O: 100 mg/mL
solubility
H2O: 100 mg/mL
Still not finding the right product?
Explore all of our products under L-Arginin
Lagerklasse
11 - Combustible Solids
Was ist los?
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Bleiben Sie mit dem Emprove®-Programm GMP-konform
Beschleunigen Sie die Qualifizierung, halten Sie die Vorschriften ein und sparen Sie Zeit mit Emprove® Dossiers.
Dossiers herunterladen
Hinweis: Sie werden zu EmproveSuite.com weitergeleitet. Ihr Benutzername und Passwort sind von SigmaAldrich.com getrennt und können abweichen.
Registrieren Sie sich kostenlos und erhalten Sie innerhalb weniger Minuten Zugang zu Emprove® Dossiers. Aktualisieren Sie Ihr Abonnement, um erweiterte und angepasste Inhalte in der Emprove® Suite freizuschalten.
Sie finden nicht, was Sie suchen? Besuchen Sie die Emprove® Programm-Hilfeseite.
Die Dokumentenbibliothek aufrufen
Können Sie Ihr Produkt oder Ihre Lot-/Chargennummer nicht finden?
Produktnummern werden bei der Anzeige auf der Website mit Packungsgrößen/-menge kombiniert (Beispiel: T1503-25G). Bitte stellen Sie sicher NUR die Produktnummer in das Feld Produktnummer eingeben (Beispiel: T1503).
Beispiel:
T1503
Produktnummer
-
25G
Packungsgröße/Menge
Weitere Beispiele:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
eingeben als 1.000309185)
Haben Sie Probleme? Sie können sich jederzeit an uns wenden Technischer Dienst zur Unterstützung