Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
human blood
manufacturer/tradename
ECACC
packaging
tube of 5 μg 88052001-DNA-5UG, pkg of vial of cells 88052001-1VL
ethnicity
Japanese
gender
male
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
-140 to -196°C
General description
Biochem/physiol Actions
Cw: 702
B: 702
HLA DRB1: 01
HLA DQB1: 05
HLA DPB1: 402
Preparation Note
Other Notes
HLA A ief: 24.1
HLA C: 7
HLA B: 7
HLA B ief: 7.2
Bw4/bw6: 6
HLA Dw: 1
HLA DR: 1
HLA DQ: 1
HLA DP: 4
Disclaimer
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 88052001, IHW Number: 9001 | description 88052080, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., IHW Number: 9081 | description 88052057, IHW Number: 9057, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description IHW Number: 9286, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 94022520 |
| manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC |
| packaging tube of 5 μg 88052001-DNA-5UG, pkg of vial of cells 88052001-1VL | packaging pkg of vial of cells 88052080-1VL, tube of 5 μg 88052080-DNA-5UG | packaging pkg of vial of cells 88052057-1VL, tube of 5 μg 88052057-DNA-5UG | packaging tube of 5 μg 94022520-DNA-5UG, pkg of vial of cells 94022520-1VL |
| karyotype Not specified | karyotype Not specified | karyotype Not specified | karyotype Not specified |
| growth mode Suspension | growth mode Suspension | growth mode Suspension | growth mode Suspension |
| biological source human blood | biological source human blood | biological source human blood | biological source human blood |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?