RAB1113
Eigenschaften
rabbit
ELISA: suitable
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 6 pg/ml
wet ice
−20°C
Beschreibung
Unterschiede anzeigen
1 of 1
Sigma-Aldrich
Rabbit PDGF-BB ELISA Kit
RAB1927
Rat PDGF-AB ELISA
RAB1152
Rat PDGF-BB ELISA Kit
RAB1223
Equine PDGF-BB ELISA
for cell culture supernatants, plasma, and serum samples
for serum, plasma and cell culture supernatants
rat
rat, human
equine
inter-assay cv: <12%intra-assay cv: <10%
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 8 pg/ml
10 - Combustible liquids
Not applicable
Danger
Resp. Sens. 1
Hier finden Sie alle aktuellen Versionen:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.
Verwandte Kategorien
Enzyme-Linked Immunosorbent Assay (ELISA) Kits
Immunoassay Platform Solutions