Fortfahren mit
1A03870
USP
D-Epinephrine
United States Pharmacopeia (USP) Reference Standard
Synonym(e):
reference material
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
Empirische Formel (Hill-System):
C9H13NO3
Molekulargewicht:
183.20
UNSPSC Code:
41116100
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
powder
shelf life
at -20 °C
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  
Other Notes
Sales restrictions may apply.
USP issued SDS can be found here
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
1A111401A077201A01450
description
United States Pharmacopeia (USP) Reference Standard
description
pharmaceutical analytical impurity (PAI)
description
pharmaceutical analytical impurity (PAI)
description
Pharmaceutical Analytical Impurity (PAI)
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical analytical impurity (PAI)
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical
pharmaceutical
pharmaceutical small molecule
shelf life
at -20 °C
shelf life
-
shelf life
-
shelf life
-
format
neat
format
neat
format
neat
format
neat
form
powder
form
-
form
-
form
-
Piktogramme
Skull and crossbones
Signalwort
Danger
Gefahrencodes
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Lagerklasse
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
Was ist los?
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)