Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
O=CNC1=CC=C(OC=2C=CN=C(C2)C(=O)NC)C=C1
InChI
InChI=1S/C14H13N3O3/c1-15-14(19)13-8-12(6-7-16-13)20-11-4-2-10(3-5-11)17-9-18/h2-9H,1H3,(H,15,19)(H,17,18)
InChI key
JPEOAANKOPAKBE-UHFFFAOYSA-N
General description
Analysis Note
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description United States Pharmacopeia (USP) Reference Standard |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical primary standard |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| agency USP | agency USP | agency USP | agency USP |
Signalwort
Danger
Gefahrencodes
Sicherheitshinweise
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2 - STOT RE 1
Zielorgane
Liver,Kidney,Pancreas,Reproductive organs,Teeth,Skin,Bone,Gastrointestinal tract
Lagerklasse
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



