Merck
  • SIRT2 deacetylase regulates the activity of GSK3 isoforms independent of inhibitory phosphorylation.
SIRT2 deacetylase regulates the activity of GSK3 isoforms independent of inhibitory phosphorylation.
eLife (2018-03-06)
Mohsen Sarikhani, Sneha Mishra, Sangeeta Maity, Chaithanya Kotyada, Donald Wolfgeher, Mahesh P Gupta, Mahavir Singh, Nagalingam R Sundaresan
ZUSAMMENFASSUNG
Glycogen synthase kinase 3 (GSK3) is a critical regulator of diverse cellular functions involved in the maintenance of structure and function. Enzymatic activity of GSK3 is inhibited by N-terminal serine phosphorylation. However, alternate post-translational mechanism(s) responsible for GSK3 inactivation are not characterized. Here, we report that GSK3α and GSK3β are acetylated at Lys246 and Lys183, respectively. Molecular modeling and/or molecular dynamics simulations indicate that acetylation of GSK3 isoforms would hinder both the adenosine binding and prevent stable interactions of the negatively charged phosphates. We found that SIRT2 deacetylates GSK3β, and thus enhances its binding to ATP. Interestingly, the reduced activity of GSK3β is associated with lysine acetylation, but not with phosphorylation at Ser9 in hearts of SIRT2-deficient mice. Moreover, GSK3 is required for the anti-hypertrophic function of SIRT2 in cardiomyocytes. Overall, our study identified lysine acetylation as a novel post-translational modification regulating GSK3 activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
(−)-Isoproterenol -hydrochlorid
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Monoklonaler Anti-β-Aktin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
GenElute Plasmid Miniprep-Kit, sufficient for 70 purifications
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
GenElute HP Plasmid Midiprep Kit, sufficient for 25 purifications
Sigma-Aldrich
Monoklonaler ANTI-FLAG-Antikörper®, clone SIG1-25, ascites fluid
Sigma-Aldrich
Natriumbutyrat, CAS-Nr. 156-54-7, Calbiochem, A short chain fatty acid that has been shown to induce apoptosis in colon carcinoma cell lines by a p53-independent pathway.
Sigma-Aldrich
Anti-Sirt2 in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Actinin antibody produced in mouse, clone BM-75.2, ascites fluid
Sigma-Aldrich
Anti-p300 CT-Antikörper, Klon RW128, clone RW128, Upstate®, from mouse
Sigma-Aldrich
Anti-Phospho-GSK3(Tyr279/Tyr216)-Antikörper, Klon 5G-2F, clone 5G-2F, Upstate®, from mouse
Sigma-Aldrich
GSK-3 Inhibitor X, The GSK-3 Inhibitor X, also referenced under CAS 740841-15-0, controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-phospho-Glycogen Synthase(Ser641/Ser645) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-SIRT2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GSK3α/β Antibody, rabbit monoclonal, culture supernatant, from rabbit