Merck
  • DNA breaks and chromatin structural changes enhance the transcription of autoimmune regulator target genes.
DNA breaks and chromatin structural changes enhance the transcription of autoimmune regulator target genes.
The Journal of biological chemistry (2017-03-01)
Mithu Guha, Mario Saare, Julia Maslovskaja, Kai Kisand, Ingrid Liiv, Uku Haljasorg, Tõnis Tasa, Andres Metspalu, Lili Milani, Pärt Peterson
ZUSAMMENFASSUNG
The autoimmune regulator (AIRE) protein is the key factor in thymic negative selection of autoreactive T cells by promoting the ectopic expression of tissue-specific genes in the thymic medullary epithelium. Mutations in AIRE cause a monogenic autoimmune disease called autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy. AIRE has been shown to promote DNA breaks via its interaction with topoisomerase 2 (TOP2). In this study, we investigated topoisomerase-induced DNA breaks and chromatin structural alterations in conjunction with AIRE-dependent gene expression. Using RNA sequencing, we found that inhibition of TOP2 religation activity by etoposide in AIRE-expressing cells had a synergistic effect on genes with low expression levels. AIRE-mediated transcription was not only enhanced by TOP2 inhibition but also by the TOP1 inhibitor camptothecin. The transcriptional activation was associated with structural rearrangements in chromatin, notably the accumulation of γH2AX and the exchange of histone H1 with HMGB1 at AIRE target gene promoters. In addition, we found the transcriptional up-regulation to co-occur with the chromatin structural changes within the genomic cluster of carcinoembryonic antigen-like cellular adhesion molecule genes. Overall, our results suggest that the presence of AIRE can trigger molecular events leading to an altered chromatin landscape and the enhanced transcription of low-expressed genes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Etoposid, synthetic, 95.0-105.0%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-CTCF-Antikörper, serum, Upstate®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.