Merck
Accelerated nuclei preparation and methods for analysis of histone modifications in yeast.
Methods (San Diego, Calif.) (2006-11-15)
Kelby O Kizer, Tiaojiang Xiao, Brian D Strahl
ZUSAMMENFASSUNG
The continuing identification of new histone post-translational modifications and ongoing discovery of their roles in nuclear processes has increased the demand for quick, efficient, and precise methods for their analysis. In the budding yeast Saccharomyces cerevisiae, a variety of methods exist for the characterization of histone modifications on a global scale. However, a wide gap in preparation time and histone purity exists between the most widely used extraction methods, which include a simple whole cell extraction (WCE) and an intensive histone extraction. In this work we evaluate various published WCE buffers for their relative effectiveness in the detection of histone modifications by Western blot analysis. We also present a precise, yet time-efficient method for the detection of subtle changes in histone modification levels. Lastly, we present a protocol for the rapid small-scale purification of nuclei that improves the performance of antibodies that do not work efficiently in WCE. These new methods are ideal for the analysis of histone modifications and could be applied to the analysis and improved detection of other nuclear proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-trimethyl-Histon H3-(Lys4-)Antikörper, Upstate®, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Dimethylhiston-H3-(Lys4)-Antikörper, Upstate®, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Dimethyl-Histon H3 (Lys36) Antikörper, serum, Upstate®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.