Merck
  • Fibroblast Growth Factor 21 Drives Dynamics of Local and Systemic Stress Responses in Mitochondrial Myopathy with mtDNA Deletions.
Fibroblast Growth Factor 21 Drives Dynamics of Local and Systemic Stress Responses in Mitochondrial Myopathy with mtDNA Deletions.
Cell metabolism (2019-09-17)
Saara Forsström, Christopher B Jackson, Christopher J Carroll, Mervi Kuronen, Eija Pirinen, Swagat Pradhan, Anastasiia Marmyleva, Mari Auranen, Iida-Marja Kleine, Nahid A Khan, Anne Roivainen, Päivi Marjamäki, Heidi Liljenbäck, Liya Wang, Brendan J Battersby, Uwe Richter, Vidya Velagapudi, Joni Nikkanen, Liliya Euro, Anu Suomalainen
ZUSAMMENFASSUNG
Mitochondrial dysfunction elicits stress responses that safeguard cellular homeostasis against metabolic insults. Mitochondrial integrated stress response (ISRmt) is a major response to mitochondrial (mt)DNA expression stress (mtDNA maintenance, translation defects), but the knowledge of dynamics or interdependence of components is lacking. We report that in mitochondrial myopathy, ISRmt progresses in temporal stages and development from early to chronic and is regulated by autocrine and endocrine effects of FGF21, a metabolic hormone with pleiotropic effects. Initial disease signs induce transcriptional ISRmt (ATF5, mitochondrial one-carbon cycle, FGF21, and GDF15). The local progression to 2nd metabolic ISRmt stage (ATF3, ATF4, glucose uptake, serine biosynthesis, and transsulfuration) is FGF21 dependent. Mitochondrial unfolded protein response marks the 3rd ISRmt stage of failing tissue. Systemically, FGF21 drives weight loss and glucose preference, and modifies metabolism and respiratory chain deficiency in a specific hippocampal brain region. Our evidence indicates that FGF21 is a local and systemic messenger of mtDNA stress in mice and humans with mitochondrial disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Fetales Kälberserum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Insulin aus Rinderpankreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
Katalase aus Rinderleber, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Cytochrom c aus Pferdeherz, ≥95% (SDS-PAGE)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Fetuin from fetal bovine serum, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Ölrot O, certified by the BSC
Sigma-Aldrich
Natriumsuccinat dibasisch Hexahydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
3,3′-Diaminobenzidin, 97% (HPLC)
Sigma-Aldrich
Anti-Gliafilamentprotein(GFAP)-Antikörper, serum, Chemicon®
Sigma-Aldrich
Actinonin
Sigma-Aldrich
Anti-LONP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF3 (1)