Merck
AGO-bound mature miRNAs are oligouridylated by TUTs and subsequently degraded by DIS3L2.
Nature communications (2020-06-04)
Acong Yang, Tie-Juan Shao, Xavier Bofill-De Ros, Chuanjiang Lian, Patricia Villanueva, Lisheng Dai, Shuo Gu
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) associated with Argonaute proteins (AGOs) regulate gene expression in mammals. miRNA 3' ends are subject to frequent sequence modifications, which have been proposed to affect miRNA stability. However, the underlying mechanism is not well understood. Here, by genetic and biochemical studies as well as deep sequencing analyses, we find that AGO mutations disrupting miRNA 3' binding are sufficient to trigger extensive miRNA 3' modifications in HEK293T cells and in cancer patients. Comparing these modifications in TUT4, TUT7 and DIS3L2 knockout cells, we find that TUT7 is more robust than TUT4 in oligouridylating mature miRNAs, which in turn leads to their degradation by the DIS3L2 exonuclease. Our findings indicate a decay machinery removing AGO-associated miRNAs with an exposed 3' end. A set of endogenous miRNAs including miR-7, miR-222 and miR-769 are targeted by this machinery presumably due to target-directed miRNA degradation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Anti-FLAG® M2 Magnetperlen, affinity isolated antibody
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.