Merck
Essentiality of CENP-A Depends on Its Binding Mode to HJURP.
Cell reports (2020-11-19)
Tetsuya Hori, JingHui Cao, Kohei Nishimura, Mariko Ariyoshi, Yasuhiro Arimura, Hitoshi Kurumizaka, Tatsuo Fukagawa
ZUSAMMENFASSUNG
CENP-A incorporation is critical for centromere specification and is mediated by the chaperone HJURP. The CENP-A-targeting domain (CATD) of CENP-A specifically binds to HJURP, and this binding is conserved. However, the binding interface of CENP-A-HJURP is yet to be understood. Here, we identify the critical residues for chicken CENP-A or HJURP. The A59Q mutation in the α1-helix of chicken CENP-A causes CENP-A mis-incorporation and subsequent cell death, whereas the corresponding mutation in human CENP-A does not. We also find that W53 of HJURP, which is a contact site of A59 in CENP-A, is also essential in chicken cells. Our comprehensive analyses reveal that the affinities of HJURP to CATD differ between chickens and humans. However, the introduction of two arginine residues to the chicken HJURP αA-helix suppresses CENP-A mis-incorporation in chicken cells expressing CENP-AA59Q. Our data explain the mechanisms and evolution of CENP-A essentiality by the CENP-A-HJURP interaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Gelatine aus Schweinehaut, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
ESGRO® Rekombinantes Maus-LIF-Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Nocodazol, ≥99% (TLC), powder
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-HA in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Casein aus Kuhmilch, suitable for substrate for protein kinase (after dephosphorylation), purified powder
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Spermin, suitable for cell culture, BioReagent
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamin, ≥97%, powder
Sigma-Aldrich
L-Histidinol -dihydrochlorid, ≥98 (TLC)
Roche
5-Brom-2′-deoxyuridin, >97%, crystalline solid, pkg of 1 g
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
1,3-Propylene sulfite, 99%