Merck
  • Transferable resistance to aminoglycosides by methylation of G1405 in 16S rRNA and to hydrophilic fluoroquinolones by QepA-mediated efflux in Escherichia coli.
Transferable resistance to aminoglycosides by methylation of G1405 in 16S rRNA and to hydrophilic fluoroquinolones by QepA-mediated efflux in Escherichia coli.
Antimicrobial agents and chemotherapy (2007-05-02)
Bruno Périchon, Patrice Courvalin, Marc Galimand
ZUSAMMENFASSUNG
Plasmid pIP1206 was detected in Escherichia coli strain 1540 during the screening of clinical isolates of Enterobacteriaceae for high-level resistance to aminoglycosides. The sequence of this IncFI conjugative plasmid of ca. 100 kb was partially determined. pIP1206 carried the rmtB gene for a ribosome methyltransferase that was shown to modify the N7 position of nucleotide G1405, located in the A site of 16S rRNA. It also contained the qepA (quinolone efflux pump) gene that encodes a 14-transmembrane-segment putative efflux pump belonging to the major facilitator superfamily of proton-dependent transporters. Disruption of membrane proton potential by the efflux pump inhibitor carbonyl cyanide m-chlorophenylhydrazone in a transconjugant harboring the qepA gene resulted in elevation of norfloxacin accumulation. The transporter conferred resistance to the hydrophilic quinolones norfloxacin and ciprofloxacin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Nalidixinsäure, ≥98%
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Norfloxacin, VETRANAL®, analytical standard