Merck
Guide to Receptors and Channels (GRAC), 4th Edition.
British journal of pharmacology (2009-11-04)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Colchicin, ≥95% (HPLC), powder
Sigma-Aldrich
Adenosin, ≥99%
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Sigma-Aldrich
Spermin, ≥97%
Sigma-Aldrich
Colchicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
L-Menthol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
Nifedipin, ≥98% (HPLC), powder
Sigma-Aldrich
Adenosin, BioReagent, suitable for cell culture
Sigma-Aldrich
Salicylsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Spermin, suitable for cell culture, BioReagent
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
L-Menthol, ≥99%, FCC, FG
Sigma-Aldrich
Salicylsäure, ≥99%, FG
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Salicylsäure, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetylsalicylsäure, analytical standard
Supelco
(−)-Menthol, analytical standard
Sigma-Aldrich
2-Amino-4-picolin, 99%
Sigma-Aldrich
(−)-Menthol, ReagentPlus®, 99%
Supelco
Ibuprofen
Sigma-Aldrich
Salicylsäure, suitable for plant cell culture