Merck
  • New plasmid-mediated quinolone resistance gene, qnrC, found in a clinical isolate of Proteus mirabilis.
New plasmid-mediated quinolone resistance gene, qnrC, found in a clinical isolate of Proteus mirabilis.
Antimicrobial agents and chemotherapy (2009-03-05)
Minghua Wang, Qinglan Guo, Xiaogang Xu, Xiaoying Wang, Xinyu Ye, Shi Wu, David C Hooper, Minggui Wang
ZUSAMMENFASSUNG
Since the discovery of qnrA in 1998, two additional qnr genes, qnrB and qnrS, have been described. These three plasmid-mediated genes contribute to quinolone resistance in gram-negative pathogens worldwide. A clinical strain of Proteus mirabilis was isolated from an outpatient with a urinary tract infection and was susceptible to most antimicrobials but resistant to ampicillin, sulfamethoxazole, and trimethoprim. Plasmid pHS10, harbored by this strain, was transferred to azide-resistant Escherichia coli J53 by conjugation. A transconjugant with pHS10 had low-level quinolone resistance but was negative by PCR for the known qnr genes, aac(6')-Ib-cr and qepA. The ciprofloxacin MIC for the clinical strain and a J53/pHS10 transconjugant was 0.25 microg/ml, representing an increase of 32-fold relative to that for the recipient, J53. The plasmid was digested with HindIII, and a 4.4-kb DNA fragment containing the new gene was cloned into pUC18 and transformed into E. coli TOP10. Sequencing showed that the responsible 666-bp gene, designated qnrC, encoded a 221-amino-acid protein, QnrC, which shared 64%, 42%, 59%, and 43% amino acid identity with QnrA1, QnrB1, QnrS1, and QnrD, respectively. Upstream of qnrC there existed a new IS3 family insertion sequence, ISPmi1, which encoded a frameshifted transposase. qnrC could not be detected by PCR, however, in 2,020 strains of Enterobacteriaceae. A new quinolone resistance gene, qnrC, was thus characterized from plasmid pHS10 carried by a clinical isolate of P. mirabilis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Nalidixinsäure, ≥98%
Supelco
Sulfamethoxazol
Supelco
Sulfamethoxazol, VETRANAL®, analytical standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard