Merck
  • Defective ALC1 nucleosome remodeling confers PARPi sensitization and synthetic lethality with HRD.
Defective ALC1 nucleosome remodeling confers PARPi sensitization and synthetic lethality with HRD.
Molecular cell (2020-12-18)
Graeme Hewitt, Valerie Borel, Sandra Segura-Bayona, Tohru Takaki, Phil Ruis, Roberto Bellelli, Laura C Lehmann, Lucia Sommerova, Aleksandra Vancevska, Antonia Tomas-Loba, Kang Zhu, Christopher Cooper, Kasper Fugger, Harshil Patel, Robert Goldstone, Deborah Schneider-Luftman, Ellie Herbert, Gordon Stamp, Rachel Brough, Stephen Pettitt, Christopher J Lord, Stephen C West, Ivan Ahel, Dragana Ahel, J Ross Chapman, Sebastian Deindl, Simon J Boulton
ZUSAMMENFASSUNG
Chromatin is a barrier to efficient DNA repair, as it hinders access and processing of certain DNA lesions. ALC1/CHD1L is a nucleosome-remodeling enzyme that responds to DNA damage, but its precise function in DNA repair remains unknown. Here we report that loss of ALC1 confers sensitivity to PARP inhibitors, methyl-methanesulfonate, and uracil misincorporation, which reflects the need to remodel nucleosomes following base excision by DNA glycosylases but prior to handover to APEX1. Using CRISPR screens, we establish that ALC1 loss is synthetic lethal with homologous recombination deficiency (HRD), which we attribute to chromosome instability caused by unrepaired DNA gaps at replication forks. In the absence of ALC1 or APEX1, incomplete processing of BER intermediates results in post-replicative DNA gaps and a critical dependence on HR for repair. Hence, targeting ALC1 alone or as a PARP inhibitor sensitizer could be employed to augment existing therapeutic strategies for HRD cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Benzonase® Nuklease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Roche
cOmplete, EDTA-freier Proteasehemmer-Cocktail, Tablets provided in glass vials
Sigma-Aldrich
Doxycyclin -hyclat
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Resazurin, powder, BioReagent
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Sox9-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Methylmethansulfonat, 99%
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
3-Indolessigsäure, suitable for plant cell culture, crystalline
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Nitrosodiethylamin, liquid
Sigma-Aldrich
2′-Desoxyuridin, ≥98.5%
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Sigma-Aldrich
Anti-RAD51-Antikörper, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Phospho-ATM(Ser1981)-Antikörper, Klon 10H11.E12, clone 10H11.E12, Upstate®, from mouse
Sigma-Aldrich
Anti-ATM antibody,Mouse monoclonal, clone MAT3-4G10/8, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Chk2-Antikörper, Klon 7, clone 7, Upstate®, from mouse
Sigma-Aldrich
Anti-BRCA1 (Ab-4) Mouse mAb (SD118), liquid, clone SD118, Calbiochem®