Merck
Islet primary cilia motility controls insulin secretion.
Science advances (2022-09-24)
Jung Hoon Cho, Zipeng A Li, Lifei Zhu, Brian D Muegge, Henry F Roseman, Eun Young Lee, Toby Utterback, Louis G Woodhams, Philip V Bayly, Jing W Hughes
ZUSAMMENFASSUNG
Primary cilia are specialized cell-surface organelles that mediate sensory perception and, in contrast to motile cilia and flagella, are thought to lack motility function. Here, we show that primary cilia in human and mouse pancreatic islets exhibit movement that is required for glucose-dependent insulin secretion. Islet primary cilia contain motor proteins conserved from those found in classic motile cilia, and their three-dimensional motion is dynein-driven and dependent on adenosine 5'-triphosphate and glucose metabolism. Inhibition of cilia motion blocks beta cell calcium influx and insulin secretion. Human beta cells have enriched ciliary gene expression, and motile cilia genes are altered in type 2 diabetes. Our findings redefine primary cilia as dynamic structures having both sensory and motile function and establish that pancreatic islet cilia movement plays a regulatory role in insulin secretion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Antimycin A aus Streptomyces sp.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-acetyliertes-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 6-11B-1, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Zytoplasmatischer Dynein-Inhibitor, Ciliobrevin D, Ciliobrevin D is a cell-permeable, reversible, and specific blocker of AAA+ ATPase motor cytoplasmic dynein. Disrupts spindle pole focusing and kinetochore-microtubule attachment (~10 to 40 µM).
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Blebistatin, racemisch, InSolution, ≥97%, 50 mM in 90% DMSO, reversible inhibitor of nonmuscle myosin II
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-GAS8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-DNALI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-DNAH5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-SPEF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-KIF9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.