Merck
Cell Death Analysis in Cancer Spheroids from a Microfluidic Device.
Methods in molecular biology (Clifton, N.J.) (2022-09-11)
Julie Lafontaine, Elena Refet-Mollof, Ouafa Najyb, Thomas Gervais, Philip Wong
ZUSAMMENFASSUNG
Microfluidic technology facilitates the generation of 3D spheroids from cancer cells, a more suitable model for preclinical therapeutic studies. This system opens the possibility to test many drugs combination at a low cost. Here we describe the use of microfluidic devices for cytotoxicity evaluation on cancer spheroids for the discovery of drugs that could be used in combination with radiotherapy. Device fabrication, preparation, and seeding are also covered. Cell death arising following treatment is detected and characterized according to spheroid size, colony formation assays, and flow cytometry analysis of apoptotic marker annexin V.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.