Merck
  • Diagnosis of prostate cancer by detection of minichromosome maintenance 5 protein in urine sediments.
Diagnosis of prostate cancer by detection of minichromosome maintenance 5 protein in urine sediments.
British journal of cancer (2010-07-22)
T J Dudderidge, J D Kelly, A Wollenschlaeger, O Okoturo, T Prevost, W Robson, H Y Leung, G H Williams, K Stoeber
ZUSAMMENFASSUNG
The accuracy of prostate-specific antigen (PSA) testing in prostate cancer detection is constrained by low sensitivity and specificity. Dysregulated expression of minichromosome maintenance (Mcm) 2-7 proteins is an early event in epithelial multistep carcinogenesis and thus MCM proteins represent powerful cancer diagnostic markers. In this study we investigate Mcm5 as a urinary biomarker for prostate cancer detection. Urine was obtained from 88 men with prostate cancer and from two control groups negative for malignancy. A strictly normal cohort included 28 men with complete, normal investigations, no urinary calculi and serum PSA <2 ng ml(-1). An expanded control cohort comprised 331 men with a benign final diagnosis, regardless of PSA level. Urine was collected before and after prostate massage in the cancer patient cohort. An immunofluorometric assay was used to measure Mcm5 levels in urine sediments. The Mcm5 test detected prostate cancer with 82% sensitivity (confidence interval (CI)= 72-89%) and with a specificity ranging from 73 (CI=68-78%) to 93% (CI=76-99%). Prostate massage led to increased Mcm5 signals compared with pre-massage samples (median 3440 (interquartile range (IQR) 2280 to 5220) vs 2360 (IQR <1800 to 4360); P=0.009), and was associated with significantly increased diagnostic sensitivity (82 vs 60%; P=0.012). Urinary Mcm5 detection seems to be a simple, accurate and noninvasive method for identifying patients with prostate cancer. Large-scale prospective trials are now required to evaluate this test in diagnosis and screening.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls