Merck
  • Dynamic changes in lipid droplet-associated proteins in the "browning" of white adipose tissues.
Dynamic changes in lipid droplet-associated proteins in the "browning" of white adipose tissues.
Biochimica et biophysica acta (2013-02-05)
David Barneda, Andrea Frontini, Saverio Cinti, Mark Christian
ZUSAMMENFASSUNG
The morphological and functional differences between lipid droplets (LDs) in brown (BAT) and white (WAT) adipose tissues will largely be determined by their associated proteins. Analysing mRNA expression in mice fat depots we have found that most LD protein genes are expressed at higher levels in BAT, with the greatest differences observed for Cidea and Plin5. Prolonged cold exposure, which induces the appearance of brown-like adipocytes in mice WAT depots, was accompanied with the potentiation of the lipolytic machinery, with changes in ATGL, CGI-58 and G0S2 gene expression. However the major change detected in WAT was the enhancement of Cidea mRNA. Together with the increase in Cidec, it indicates that LD enlargement through LD-LD transference of fat is an important process during WAT browning. To study the dynamics of this phenotypic change, we have applied 4D confocal microscopy in differentiated 3T3-L1 cells under sustained β-adrenergic stimulation. Under these conditions the cells experienced a LD remodelling cycle, with progressive reduction on the LD size by lipolysis, followed by the formation of new LDs, which were subjected to an enlargement process, likely to be CIDE-triggered, until the cell returned to the basal state. This transformation would be triggered by the activation of a thermogenic futile cycle of lipolysis/lipogenesis and could facilitate the molecular mechanism for the unilocular to multilocular transformation during WAT browning. This article is part of a Special Issue entitled Brown and White Fat: From Signaling to Disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized powder, ≥15 units/mg solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Esterase aus Schweineleber, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized, powder, slightly beige, ≥50 U/mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Esterase aus Kaninchenleber, lyophilized powder, ≥30 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Esteraseisoenzym-1 aus Schweineleber, rekombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Preise und Verfügbarkeit sind derzeit nicht verfügbar.