Merck
  • Design, synthesis, and structure-activity relationship studies of novel 3-alkylindole derivatives as selective and highly potent myeloperoxidase inhibitors.
Design, synthesis, and structure-activity relationship studies of novel 3-alkylindole derivatives as selective and highly potent myeloperoxidase inhibitors.
Journal of medicinal chemistry (2013-04-16)
Jalal Soubhye, Iyas Aldib, Betina Elfving, Michel Gelbcke, Paul G Furtmüller, Manuel Podrecca, Raphaël Conotte, Jean-Marie Colet, Alexandre Rousseau, Florence Reye, Ahmad Sarakbi, Michel Vanhaeverbeek, Jean-Michel Kauffmann, Christian Obinger, Jean Nève, Martine Prévost, Karim Zouaoui Boudjeltia, Francois Dufrasne, Pierre Van Antwerpen
ZUSAMMENFASSUNG
Due to its production of potent antimicrobial oxidants including hypochlorous acid, human myeloperoxidase (MPO) plays a critical role in innate immunity and inflammatory diseases. Thus MPO is an attractive target in drug design. (Aminoalkyl)fluoroindole derivatives were detected to be very potent MPO inhibitors; however, they also promote inhibition of the serotonin reuptake transporter (SERT) at the same concentration range. Via structure-based drug design, a new series of MPO inhibitors derived from 3-alkylindole were synthesized and their effects were assessed on MPO-mediated taurine chlorination and low-density lipoprotein oxidation as well as on inhibition of SERT. The fluoroindole compound with three carbons in the side chain and one amide group exhibited a selectivity index of 35 (Ki/IC50) with high inhibition of MPO activity (IC50 = 18 nM), whereas its effect on SERT was in the micromolar range. Structure-function relationships, mechanism of action, and safety of the molecule are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Peroxidase aus Meerrettich, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Peroxidase aus Meerrettich, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Myeloperoxidase aus Humanleukocyten, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Taurin, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Peroxidase aus Meerrettich, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Lactoperoxidase aus Kuhmilch, lyophilized powder (essentially salt-free), ≥200 units/mg protein
Supelco
Taurin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurin, ≥98%, FG
Sigma-Aldrich
Peroxidase aus Meerrettich, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase aus Meerrettich, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Schwefel, flakes, ≥99.99% trace metals basis
SAFC
Taurin
Supelco
Schwefel, PESTANAL®, analytical standard
Sigma-Aldrich
Lactoperoxidase aus Kuhmilch, lyophilized, powder, ≥150 U/mg