Merck
  • A probability-based approach for high-throughput protein phosphorylation analysis and site localization.
A probability-based approach for high-throughput protein phosphorylation analysis and site localization.
Nature biotechnology (2006-09-12)
Sean A Beausoleil, Judit Villén, Scott A Gerber, John Rush, Steven P Gygi
ZUSAMMENFASSUNG
Data analysis and interpretation remain major logistical challenges when attempting to identify large numbers of protein phosphorylation sites by nanoscale reverse-phase liquid chromatography/tandem mass spectrometry (LC-MS/MS) (Supplementary Figure 1 online). In this report we address challenges that are often only addressable by laborious manual validation, including data set error, data set sensitivity and phosphorylation site localization. We provide a large-scale phosphorylation data set with a measured error rate as determined by the target-decoy approach, we demonstrate an approach to maximize data set sensitivity by efficiently distracting incorrect peptide spectral matches (PSMs), and we present a probability-based score, the Ascore, that measures the probability of correct phosphorylation site localization based on the presence and intensity of site-determining ions in MS/MS spectra. We applied our methods in a fully automated fashion to nocodazole-arrested HeLa cell lysate where we identified 1,761 nonredundant phosphorylation sites from 491 proteins with a peptide false-positive rate of 1.3%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phospholipase A2 aus Honigbienengift (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Phospholipase A2 aus Schweinepankreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 aus Rinderpankreas, lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein