Merck
  • Tunable pH-sensitive poly(β-amino ester)s synthesized from primary amines and diacrylates for intracellular drug delivery.
Tunable pH-sensitive poly(β-amino ester)s synthesized from primary amines and diacrylates for intracellular drug delivery.
Macromolecular bioscience (2012-08-29)
Wantong Song, Zhaohui Tang, Mingqiang Li, Shixian Lv, Haiyang Yu, Lili Ma, Xiuli Zhuang, Yubin Huang, Xuesi Chen
ZUSAMMENFASSUNG
The pH sensitivity of a series of PbAEs synthesized from primary amines and diacrylates is studied. By changing alkyl groups of the amine monomers, the pKb can be tuned across a broad range (from 3.5 to 7.2). Micelles formed from a PEG-PbAE block copolymer retain the pH sensitivity of PbAE and can stably load hydrophobic molecules under neutral pH, while quickly dissociate and release their cargoes at pH ≈ 6.0. When the chemotherapy drug DOX is loaded, the micelles show efficient cell proliferation inhibition to HeLa cells and fast intracellular release. Thus, the primary-amine-based PbAEs are shown to be promising in the construction of intracellular targeting drug delivery systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nilrot, suitable for microscopy
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nilrot, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nilrot, Technical grade
Preise und Verfügbarkeit sind derzeit nicht verfügbar.