Merck
  • A photoactive basket-like metal-organic tetragon worked as an enzymatic molecular flask for light driven H2 production.
A photoactive basket-like metal-organic tetragon worked as an enzymatic molecular flask for light driven H2 production.
Chemical communications (Cambridge, England) (2012-12-12)
Cheng He, Jian Wang, Liang Zhao, Tao Liu, Jing Zhang, Chunying Duan
ZUSAMMENFASSUNG
A photoactive basket-like metal-organic tetragon Ce-ZL that contained a carbazole photosensitizer was developed to capture the biomimetic [FeFe]-H(2)ases for light driven H(2) production. The system exhibited enzymatic behaviour and its activity was inhibited by the encapsulation of ATP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carbazol, ≥95% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Carbazol, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.