Merck
  • The hydrogen-bonded dianion of vitamin K1 produced in aqueous-organic solutions exists in equilibrium with its hydrogen-bonded semiquinone anion radical.
The hydrogen-bonded dianion of vitamin K1 produced in aqueous-organic solutions exists in equilibrium with its hydrogen-bonded semiquinone anion radical.
The journal of physical chemistry. B (2013-02-13)
Zhen Hui Lim, Elaine Lay Khim Chng, Yanlan Hui, Richard D Webster
ZUSAMMENFASSUNG
When the quinone, vitamin K1 (VK1), is electrochemically reduced in aqueous-acetonitrile solutions (CH3CN with 7.22 M H2O), it undergoes a two-electron reduction to form the dianion that is hydrogen-bonded with water [VK1(H2O)y(2–)]. EPR and voltammetry experiments have shown that the persistent existence of the semiquinone anion radical (also hydrogen-bonded with water) [VK1(H2O)x(–•)] in aqueous or organic–aqueous solutions is a result of VK1(H2O)y(2–) undergoing a net homogeneous electron transfer reaction (comproportionation) with VK1, and not via direct one-electron reduction of VK1. When 1 mM solutions of VK1 were electrochemically reduced by two electrons in aqueous-acetonitrile solutions, quantitative EPR experiments indicated that the amount of VK1(H2O)x(–•) produced was up to approximately 35% of all the reduced species. In situ electrochemical ATR-FTIR experiments on sequentially one- and two-electron bulk reduced solutions of VK1 (showing strong absorbances at 1664, 1598, and 1298 cm(–1)) in CH3CN containing <0.05 M H2O led to the detection of VK1(–•) with strong absorbances at 1710, 1703, 1593, 1559, 1492, and 1466 cm(–1) and VK1(H2O)y(2–) with strong absorbances at 1372 and 1342 cm(–1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Phyllochinon (K1), analytical standard
Supelco
Phytonadion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Phytomenadion, European Pharmacopoeia (EP) Reference Standard