Merck
  • Novel ternary vanadium-betaine-peroxido species suppresses H-ras and matrix metalloproteinase-2 expression by increasing reactive oxygen species-mediated apoptosis in cancer cells.
Novel ternary vanadium-betaine-peroxido species suppresses H-ras and matrix metalloproteinase-2 expression by increasing reactive oxygen species-mediated apoptosis in cancer cells.
Cancer letters (2013-03-12)
Savvas Petanidis, Efrosini Kioseoglou, Margarita Hadzopoulou-Cladaras, Athanasios Salifoglou
ZUSAMMENFASSUNG
Vanadium is known for its antitumorigenicity. Poised to investigate the impact of well-defined forms of vanadium on processes and specific biomolecules (oncogenes-proteins) involved in cancer cell physiology, a novel ternary V(V)-peroxido-betaine compound was employed in experiments targeting cell viability, apoptosis, reactive oxygen species (ROS) production, H-ras signaling, and matrix metalloproteinase-2 (MMP-2) expression in human breast cancer epithelial and lung adenocarcinoma cells. The results reveal that vanadium imparts a significant decrease in cancer cell viability, reducing H-ras and MMP-2 expression by increasing ROS-mediated apoptosis, distinctly emphasizing the nature, structure and properties of ternary ligands on vanadium anti-tumor activity and its future potential as a metallodrug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Betain -Lösung, 5 M, PCR Reagent
Sigma-Aldrich
Betain, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betain, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betain -hydrochlorid, ≥99%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis