Merck
Curcumin inhibits TGFβ1-induced CCN2 via Src, JNK, and Smad3 in gingiva.
Journal of dental research (2013-04-24)
W-H Yang, M Y-P Kuo, C-M Liu, Y-T Deng, H-H Chang, J Z-C Chang
ZUSAMMENFASSUNG
Transforming growth factor β (TGFβ) is a key regulator associated with the pathogenesis of gingival overgrowth (GO). Connective tissue growth factor (CTGF/CCN2) is overexpressed in GO tissues. CCN2 promotes and sustains fibrosis initiated by TGFβ. Previous studies have shown that JNK and Smad3 activation is required for TGFβ-induced CCN2 expressions in human gingival fibroblasts (HGFs). In this study, we have found that Src is a major signaling mediator for TGFβ-induced CCN2 expressions in HGFs. Pre-treatment with 2 Src kinase inhibitors (PP2, Src inhibitor-1) significantly reduced TGFβ1-induced CCN2 synthesis and JNK and Smad3 activation in HGFs. These results suggest that Src is an upstream signaling transducer of JNK and Smad3 with respect to TGFβ1-stimulated CCN2 expression in HGFs. We further found that curcumin significantly abrogated the TGFβ1-induced CCN2 in HGFs by inhibiting the phosphorylations of Src, JNK, and Smad3. Furthermore, curcumin inhibited TGFβ1-induced HGF migration and α-SMA expression. Curcumin potentially qualifies as a useful agent for the control of GO.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥95%
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥80% (HPLC), from green tea
Supelco
Curcumin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Supelco
(+)-Catechin, analytical standard
Supelco
(−)-Epigallokatechingallat, analytical standard
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Epicatechin, primary reference standard
Supelco
(−)-Epicatechin, analytical standard
Epigallocatechin-gallat, primary reference standard
Curcumin, primary reference standard
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Lovastatin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard