Merck
Plasma amino acid profile on nocturnal hemodialysis.
Blood purification (2000-06-06)
D S Raj, M Ouwendyk, R Francoeur, A Pierratos
ZUSAMMENFASSUNG
The purpose of this study was to examine the effect of nocturnal hemodialysis (NHD) on serum amino acid (AA) profile. In a cross-over trial, we studied the AA profile in 11 patients who were switched from conventional hemodialysis to NHD. Plasma levels of AA were measured at monthly intervals for a period of 1 year. Concentrations of AA isoleucine, threonine, phenylalanine, valine, glycine, glutamine and taurine increased on switch to NHD. Total AA (2,907.2 +/- 542.2 vs. 3,870.2 +/- 715.2 micromol/l, p = 0.03), essential AA (EAA, 842.2 +/- 259.4 vs. 1,125. 8 +/- 235.3 micromol/l, p = 0.03), nonessential AA (NEAA, 2,307.2 +/- 399.1 vs. 2,879.5 +/- 581.2 micromol/l, p = 0.03), and branched chain AA (315.5 +/- 90.8 vs. 386.0 +/- 114.3 micromol/l, p = 0.05) concentrations were significantly higher on NHD. However, abnormalities in the ratios of EAA/NEAA, valine/glycine and tyrosine/phenylalanine persisted during NHD. NHD alleviated some abnormalities in the aminogram of uremia, but other metabolic derangements lingered.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Betain -Lösung, 5 M, PCR Reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Betain, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Glycolsäure, ReagentPlus®, 99%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Betain, ≥98% (perchloric acid titration)
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
p-Cresol, 99%
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
SAFC
L-Prolin
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material