Merck
Dimethylglycine accumulates in uremia and predicts elevated plasma homocysteine concentrations.
Kidney international (2001-05-31)
D O McGregor, W J Dellow, M Lever, P M George, R A Robson, S T Chambers
ZUSAMMENFASSUNG
Hyperhomocysteinemia is a risk factor for atherosclerosis that is common in chronic renal failure (CRF), but its cause is unknown. Homocysteine metabolism is linked to betaine-homocysteine methyl transferase (BHMT), a zinc metalloenzyme that converts glycine betaine (GB) to N,N dimethylglycine (DMG). DMG is a known feedback inhibitor of BHMT. We postulated that DMG might accumulate in CRF and contribute to hyperhomocysteinemia by inhibiting BHMT activity. Plasma and urine concentrations of GB and DMG were measured in 33 dialysis patients (15 continuous ambulatory peritoneal dialysis and 18 hemodialysis), 33 patients with CRF, and 33 age-matched controls. Concentrations of fasting plasma total homocysteine (tHcy), red cell and serum folate, vitamins B(6) and B(12), serum zinc, and routine biochemistry were also measured. Groups were compared, and determinants of plasma tHcy were identified by correlations and stepwise linear regression. Plasma DMG increased as renal function declined and was twofold to threefold elevated in dialysis patients. Plasma GB did not differ between groups. The fractional excretion of GB (FE(GB)) was increased tenfold, and FED(MG) was doubled in CRF patients compared with controls. Plasma tHcy correlated positively with plasma DMG, the plasma DMG:GB ratio, plasma creatinine, and FE(GB) and negatively with serum folate, zinc, and plasma GB. In the multiple regression model, only plasma creatinine, plasma DMG, or the DMG:GB ratio was independent predictors of tHcy. DMG accumulates in CRF and independently predicts plasma tHcy concentrations. These findings suggest that reduced BHMT activity is important in the pathogenesis of hyperhomocysteinemia in CRF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Betain -Lösung, 5 M, PCR Reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Betain, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Glycolsäure, ReagentPlus®, 99%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Betain, ≥98% (perchloric acid titration)
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
p-Cresol, 99%
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
SAFC
L-Prolin
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material