Merck
Metal presence in hair after metal-on-metal resurfacing arthroplasty.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2013-07-31)
María Rodríguez de la Flor, Daniel Hernández-Vaquero, José Manuel Fernández-Carreira
ZUSAMMENFASSUNG
The elevation of metal levels in serum and urine during post-operative follow-up is a frequent find following the implantation of certain models of metal-on-metal hip prostheses. Among 45 patients with the same resurfacing prostheses, chromium (Cr) and cobalt (Co) levels in serum and urine were determined at 3, 4, 5, and 6 years after surgery. In the same period, levels of Cr, Co and molibdene (Mo) in scalp hair were also measured. Mean Cr and Co levels in serum were 8.29 µg/L (SD 17.97) and 8.38 µg/L (SD 21.97), respectively, whereas in urine levels were 16.20 µg/L (SD SD 32.55) and 75.40 µg/L (SD 190.86), respectively. In hair, mean Cr level were 163.27 µg/g (SD 300.62), mean Co level 61.98 µg/g (SD 126.48), and Mo 31.36 µg/g (SD 37.86). A high concordance was observed between chromium-urine and chromium-serum and between cobalt-urine and cobalt-hair. A moderate concordance was present between cobalt-urine and cobalt-serum, and between cobalt-hair and cobalt-serum. Eleven patients required revision surgery, five of them due to metallosis and periarticular cyst. At 1 year after reintervention, analytics were performed again and the following decrease rates were found: 42.8% in Cr levels, 51.1% in Mo levels, and 90.3% in Co levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Molybdän, powder, <150 μm, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, powder, 10 μm, ≥99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, wire, diam. 1.0 mm, 99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Molybdän, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.