Merck
  • Identification and characterization of the related immune-enhancing proteins in crab Scylla paramamosain stimulated with rhubarb polysaccharides.
Identification and characterization of the related immune-enhancing proteins in crab Scylla paramamosain stimulated with rhubarb polysaccharides.
Molecular immunology (2013-11-12)
Jingsong Cao, Zehuan Wang, Yueling Zhang, Fengliang Qu, Lingling Guo, Mingqi Zhong, Shengkang Li, Haiying Zou, Jiehui Chen, Xiuying Wang
ZUSAMMENFASSUNG
Recently, considerable interest has been focused on immunostimulants to reduce diseases in crab aquaculture. However, information regarding to the related immune-enhancing proteins in crabs is not available yet. In this study, rhubarb polysaccharides were tested for enhancement of the immune activity in crab Scylla paramamosain. Compared with those in the control group, values of, phenoloxidase (PO), alkaline phosphatase (AKP) and alkaline phosphatasein (ACP) activity in the, experimental group were improved significantly 4 d after the treatment. Furthermore, 15 and 17 altered proteins from haemocytes and hepatopancreas, respectively, were found in rhubarb polysaccharide-treated crabs using 2-DE approach. Of these, hemocyanin, chymotrypsin, cryptocyanin, C-type lectin receptor, and ferritin protein were identified by mass spectrometry. In addition, RT-PCR, analysis showed that the mRNA levels of hemocyanin and chymotrypsin increased about 2.4- and 1.4-fold in the experiment group. Moreover, the hemocyanin gene in S. paramamosain (SpHMC) was, cloned and characterized. SpHMC contains one open reading frame of 2022 bp and encodes a polypeptide of 673 amino acids. It is clustered into one branch along with crab hemocyanin in a phylogenetic tree. The mRNA transcripts of SpHMC were detected mainly in the tissues of, hepatopancreas, hemocyte and intestines, and its levels were up-regulated significantly in hemocytes, of S. paramamosain treated with Vibrio parahemolyticus, Beta streptococcus or poly I:C for 6-48 h. Taken together, these studies found 5 related immune-enhancing proteins and a novel heomcyanin homologue with potential pathogen-resistant activities in crab.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyrosinase aus Ständerpilz, lyophilized powder, ≥1000 unit/mg solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type II, lyophilized powder, ≥40 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
αα-Chymotrypsin, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type I-S, essentially salt-free, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, ≥40 units/mg protein, vial of 5 mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, suitable for protein sequencing, salt-free, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Schweineniere, lyophilized powder, ≥100 DEA units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch aus Kälberdarmschleimhaut, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalische Garnelen, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphatase, alkalisch Rind, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.