Merck
The fibrillogenic L178H variant of apolipoprotein A-I forms helical fibrils.
Journal of lipid research (2011-12-21)
Jitka Petrlova, Trang Duong, Megan C Cochran, Annika Axelsson, Matthias Mörgelin, Linda M Roberts, Jens O Lagerstedt
ZUSAMMENFASSUNG
A number of amyloidogenic variants of apoA-I have been discovered but most have not been analyzed. Previously, we showed that the G26R mutation of apoA-I leads to increased β-strand structure, increased N-terminal protease susceptibility, and increased fibril formation after several days of incubation. In vivo, this and other variants mutated in the N-terminal domain (residues 26 to ∼90) lead to renal and hepatic accumulation. In contrast, several mutations identified within residues 170 to 178 lead to cardiac, laryngeal, and cutaneous protein deposition. Here, we describe the structural changes in the fibrillogenic variant L178H. Like G26R, the initial structure of the protein exhibits altered tertiary conformation relative to wild-type protein along with decreased stability and an altered lipid binding profile. However, in contrast to G26R, L178H undergoes an increase in helical structure upon incubation at 37°C with a half time (t(1/2)) of about 12 days. Upon prolonged incubation, the L178H mutant forms fibrils of a diameter of 10 nm that ranges in length from 30 to 120 nm. These results show that apoA-I, known for its dynamic properties, has the ability to form multiple fibrillar conformations, which may play a role in the tissue-specific deposition of the individual variants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktailpulver, for general use, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteasehemmer-Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.