Merck
  • Efficacy of sugammadex for the reversal of moderate and deep rocuronium-induced neuromuscular block in patients pretreated with intravenous magnesium: a randomized controlled trial.
Efficacy of sugammadex for the reversal of moderate and deep rocuronium-induced neuromuscular block in patients pretreated with intravenous magnesium: a randomized controlled trial.
Anesthesiology (2014-03-13)
Christoph Czarnetzki, Edömér Tassonyi, Christopher Lysakowski, Nadia Elia, Martin R Tramèr
ZUSAMMENFASSUNG
Magnesium enhances the effect of rocuronium. Sugammadex reverses rocuronium-induced neuromuscular block. The authors investigated whether magnesium decreased the efficacy of sugammadex for the reversal of rocuronium-induced neuromuscular block. Thirty-two male patients were randomized in a double-blinded manner to receive magnesium sulfate (MgSO4) 60 mg/kg or placebo intravenously before induction of anesthesia with propofol, sufentanil, and rocuronium 0.6 mg/kg. Neuromuscular transmission was monitored using TOF-Watch SX acceleromyography (Organon Ltd., Dublin, Ireland). In 16 patients, sugammadex 2 mg/kg was administered intravenously at reappearance of the second twitch of the train-of-four (moderate block). In 16 further patients, sugammadex 4 mg/kg was administered intravenously at posttetanic count 1 to 2 (deep block). Primary endpoint was recovery time from injection of sugammadex to normalized train-of-four ratio 0.9. Secondary endpoint was recovery time to final T1. Average time for reversal of moderate block was 1.69 min (SD, 0.81) in patients pretreated with MgSO4 and 1.76 min (1.13) in those pretreated with placebo (P = 0.897). Average time for reversal of deep block was 1.77 min (0.83) in patients pretreated with MgSO4 and 1.98 min (0.58) in those pretreated with placebo (P = 0.572). Times to final T1 were longer compared with times to normalized train-of-four ratio 0.9, without any difference between patients pretreated with MgSO4 or placebo. Pretreatment with a single intravenous dose of MgSO4 60 mg/kg does not decrease the efficacy of recommended doses of sugammadex for the reversal of a moderate and deep neuromuscular block induced by an intubation dose of rocuronium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat Heptahydrat, ACS reagent, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat Heptahydrat, ReagentPlus®, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat Heptahydrat, BioReagent, Molecular Biology, suitable for plant cell culture, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat Heptahydrat, BioUltra, ≥99.5% (KT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat -Lösung, BioUltra, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat Heptahydrat, BioXtra, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Rocuronium -bromid, ≥97% (perchloric acid titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Rocuronium für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Rocuronium -bromid, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.