Merck
New insight into active muscarinic receptors with the novel radioagonist [³H]iperoxo.
Biochemical pharmacology (2014-05-28)
Ramona Schrage, Janine Holze, Jessica Klöckner, Aileen Balkow, Anne S Klause, Anna-Lena Schmitz, Marco De Amici, Evi Kostenis, Christian Tränkle, Ulrike Holzgrabe, Klaus Mohr
ZUSAMMENFASSUNG
Activation of G protein-coupled receptors involves major conformational changes of the receptor protein ranging from the extracellular transmitter binding site to the intracellular G protein binding surface. GPCRs such as the muscarinic acetylcholine receptors are commonly probed with radioantagonists rather than radioagonists due to better physicochemical stability, higher affinity, and indifference towards receptor coupling states of the former. Here we introduce tritiated iperoxo, a superagonist at muscarinic M₂ receptors with very high affinity. In membrane suspensions of transfected CHO-cells, [³H]iperoxo - unlike the common radioagonists [³H]acetylcholine and [³H]oxotremorine M - allowed labelling of each of the five muscarinic receptor subtypes in radioagonist displacement and saturation binding studies. [³H]iperoxo revealed considerable differences in affinity between the even- and the odd-numbered muscarinic receptor subtypes with affinities for the M₂ and M₄ receptor in the picomolar range. Probing ternary complex formation on the M₂ receptor, [³H]iperoxo dissociation was not influenced by an archetypal allosteric inverse agonist, reflecting activation-related rearrangement of the extracellular loop region. At the inner side of M₂, the preferred Gi protein acted as a positive allosteric modulator of [³H]iperoxo binding, whereas Gs and Gq were neutral in spite of their robust coupling to the activated receptor. In intact CHO-hM₂ cells, endogenous guanylnucleotides promoted receptor/G protein-dissociation resulting in low-affinity agonist binding which, nevertheless, was still reported by [³H]iperoxo. Taken together, the muscarinic superagonist [³H]iperoxo is the best tool currently available for direct probing activation-related conformational transitions of muscarinic receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(−)-N-Methylhyosciniumbromid, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Oxitropiumbromid Unreinheit B, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.