Merck
Methotrexate for induction of remission in refractory Crohn's disease.
The Cochrane database of systematic reviews (2014-08-08)
John W D McDonald, Yongjun Wang, David J Tsoulis, John K MacDonald, Brian G Feagan
ZUSAMMENFASSUNG
Although corticosteroids are effective for induction of remission of Crohn's disease, many patients relapse when steroids are withdrawn or become steroid dependent. Furthermore, corticosteroids exhibit significant adverse effects. The success of methotrexate as a treatment for rheumatoid arthritis led to its evaluation in patients with refractory Crohn's disease. Methotrexate has been studied for induction of remission of refractory Crohn's disease and has become the principal alternative to azathioprine or 6-mercaptopurine therapy. This systematic review is an update of previously published Cochrane reviews. The primary objective was to assess the efficacy and safety of methotrexate for induction of remission in patients with active Crohn's disease in the presence or absence of concomitant steroid therapy. We searched MEDLINE, EMBASE, CENTRAL and the Cochrane IBD/FBD group specialized register from inception to June 9, 2014 for relevant studies. Conference proceedings and reference lists were also searched to identify additional studies. Randomized controlled trials of methotrexate compared to placebo or an active comparator for treatment of active refractory Crohn's disease in adult patients (> 17 years) were considered for inclusion. The primary outcome was failure to enter remission and withdraw from steroids. Secondary outcomes included adverse events, withdrawal due to adverse events, serious adverse events and quality of life. We calculated the relative risk (RR) and 95% confidence intervals (95% CI) for each outcome. Data were analyzed on an intention-to-treat basis. The Cochrane risk of bias tool was used to assess the methodological quality of included studies. The GRADE approach was used to assess the overall quality of evidence supporting the primary outcome. Seven studies (495 patients) were included. Four studies were rated as low risk of bias. Three studies were rated as high risk of bias due to open label or single-blind designs. The seven studies differed with respect to participants, intervention, and outcomes to the extent that meta-analysis was considered to be inappropriate. GRADE analyses indicated that the quality of evidence was very low to low for most outcomes due to sparse data and inadequate blinding. Three small studies which employed low dose oral methotrexate showed no statistically significant difference in failure to induce remission between methotrexate and placebo or between methotrexate and 6-mercaptopurine. For the study using 15 mg/week of oral methotrexate 33% (5/15) of methotrexate patients failed to enter remission compared to 11% (2/18) of placebo patients (RR 3.00, 95% CI 0.68 to 13.31). For the study using 12.5 mg/week of oral methotrexate 81% (21/26) of methotrexate patients failed to enter remission compared to 77% (20/26) of placebo patients (RR 1.05, 95% CI 0.79 to 1.39). This study also had an active comparator arm, 81% (21/26) of methotrexate patients failed to enter remission compared to 59% (19/32) of 6-mercaptopurine patients (RR 1.36, 95% CI 0.97 to 1.92). For the active comparator study using 15 mg/week oral methotrexate, 20% (3/15) of methotrexate patients failed to enter remission compared to 6% of 6-mercaptopurine patients (RR 3.20, 95% CI 0.37 to 27.49). This study also had a 5-ASA arm and found that methotrexate patients were significantly more likely to enter remission than 5-ASA patients. Twenty per cent (3/15) of methotrexate patients failed to enter remission compared to 86% (6/7) of 5-ASA patients (RR 0.23, 95% CI 0.08 to 0.67). One small study which used a higher dose of intravenous or oral methotrexate (25 mg/week) showed no statistically significant difference between methotrexate and azathioprine. Forty-four per cent (12/27) of methotrexate patients failed to enter remission compared to 37% of azathioprine patients (RR 1.20, 95% CI 0.63 to 2.29). Two studies found no statistically significant difference in failure to enter remission between the combination of infliximab and methotrexate and infliximab monotherapy. One small study utilized intravenous methotrexate (20 mg/week) for 5 weeks and then switched to oral (20 mg/week). Forty-five per cent (5/11) of patients in the combination group failed to enter remission compared to 62% of infliximab patients (RR 0.73, 95% CI 0.31 to 1.69). The other study assessing combination therapy utilized subcutaneous methotrexate (maximum dose 25 mg/week). Twenty-four per cent (15/63) of patients in the combination group failed to enter remission compared to 22% (14/63) of infliximab patients (RR 1.07, 95% CI 0.57 to 2.03). A large placebo-controlled study which employed a high dose of methotrexate intramuscularly showed a statistically significant benefit relative to placebo. Sixty-one per cent of methotrexate patients failed to enter remission compared to 81% of placebo patients (RR 0.75, 95% CI 0.61 to 0.93; number needed to treat, NNT=5). Withdrawals due to adverse events were significantly more common in methotrexate patients than placebo in this study. Seventeen per cent of methotrexate patients withdrew due to adverse events compared to 2% of placebo patients (RR 8.00, 95% CI 1.09 to 58.51). The incidence of adverse events was significantly more common in methotrexate patients (63%, 17/27) than azathioprine patients (26%, 7/27) in one small study (RR 2.42, 95% CI 1.21 to 4.89). No other statistically significant differences in adverse events, withdrawals due to adverse events or serious adverse events were reported in any of the other placebo-controlled or active comparator studies. Common adverse events included nausea and vomiting, abdominal pain, diarrhea, skin rash and headache. There is evidence from a single large randomized trial which suggests that intramuscular methotrexate (25 mg/week) provides a benefit for induction of remission and complete withdrawal from steroids in patients with refractory Crohn's disease. Lower dose oral methotrexate does not appear to provide any significant benefit relative to placebo or active comparator. However, these trials were small and further studies of oral methotrexate may be justified. Comparative studies of methotrexate to drugs such as azathioprine or 6-mercaptopurine would require the randomization of large numbers of patients. The addition of methotrexate to infliximab therapy does not appear to provide any additional benefit over infliximab monotherapy. However these studies were relatively small and further research is needed to determine the role of methotrexate when used in conjunction with infliximab or other biological therapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Prednison, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Amethopterin Hydrat, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Prednison, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Amethopterin Hydrat, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methotrexat, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Methotrexat
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methotrexat, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Prednison, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
6-Mercaptopurin Monohydrat, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methotrexat -Lösung, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Azathioprin, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Methotrexat, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Amethopterin Hydrat, ≥99.0% (sum of enantiomers, HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Azathioprin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Azathioprin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Methotrexat für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Methotrexat für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.