Merck
  • Nickel-catalyzed intramolecular [3 + 2 + 2] cycloadditions of alkylidenecyclopropanes. A straightforward entry to fused 6,7,5-tricyclic systems.
Nickel-catalyzed intramolecular [3 + 2 + 2] cycloadditions of alkylidenecyclopropanes. A straightforward entry to fused 6,7,5-tricyclic systems.
Organic letters (2014-09-19)
Lucía Saya, Israel Fernández, Fernando López, José L Mascareñas
ZUSAMMENFASSUNG
A highly diastereo- and chemoselective intramolecular nickel-catalyzed cycloaddition of alkene- and alkyne-tethered alkynylidenecyclopropanes is reported. The method constitutes the first fully intramolecular [3 + 2 + 2] alkylidenecyclopropropane cycloaddition occurring via a proximal cleavage of the cyclopropane and makes it possible to build relevant 6,7,5-tricyclic frameworks in a single-pot reaction. Importantly, the reaction outcome is highly dependent on the characteristics of the nickel ligands.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Nickel, rod, 100mm, diameter 1.5mm, 99.99+%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, pellets, 100g, max. size 10mm, 99.999%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Nickel, mesh, 30x30mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Nickel, foil, 100x100mm, thickness 1.6mm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Nickel, foil, light tested, 150x150mm, thickness 0.025mm, 99.9%
Nickel, mesh, 100x250mm, nominal aperture 0.73mm, wire diameter 0.25mm, 26x26 wires/inch, open area 55%, plain weave mesh, 99%
Nickel, pellets, 100g, max. size 5mm, 99.99+%
Nickel, wire reel, 100m, diameter 0.025mm, as drawn, 99.98%
Nickel, mesh, 65x65mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Nickel, foil, 100x100mm, thickness 0.1mm, annealed, 99%
Nickel, mesh, 130x130mm, nominal aperture 0.04mm, thickness 0.004mm, wire diameter 0.011mm, 500 wires/inch, open area 60%, electro-formed mesh, 99.9%
Nickel, sphere, 100pcs, diameter 1.0mm, precision sphere grade 200, 99%
Nickel, mesh, 120x120mm, nominal aperture 0.43mm, thickness 0.004mm, wire diameter 0.067mm, 51 wires/inch, open area 75%, electro-formed mesh, 99.9%