Merck
  • Optimization of radiation therapy techniques for prostate cancer with prostate-rectum spacers: a systematic review.
Optimization of radiation therapy techniques for prostate cancer with prostate-rectum spacers: a systematic review.
International journal of radiation oncology, biology, physics (2014-10-12)
Gary Mok, Eileen Benz, Jean-Paul Vallee, Raymond Miralbell, Thomas Zilli
ZUSAMMENFASSUNG
Dose-escalated radiation therapy for localized prostate cancer improves disease control but is also associated with worse rectal toxicity. A spacer placed between the prostate and rectum can be used to displace the anterior rectal wall outside of the high-dose radiation regions and potentially minimize radiation-induced rectal toxicity. This systematic review focuses on the published data regarding the different types of commercially available prostate-rectum spacers. Dosimetric results and preliminary clinical data using prostate-rectum spacers in patients with localized prostate cancer treated by curative radiation therapy are compared and discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Rinderaugen-Glaskörper
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 8,000-15,000
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,500,000-1,750,000
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 130,000-150,000
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,200
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 300,000-500,000