Merck
Temporal trends in organic carbon content in the main Swiss rivers, 1974-2010.
The Science of the total environment (2014-09-27)
J C Rodríguez-Murillo, J Zobrist, M Filella
ZUSAMMENFASSUNG
Increases in dissolved organic carbon (DOC) concentrations have often been reported in rivers and lakes of the Northern Hemisphere over the last few decades. High-quality organic carbon (OC) concentration data have been used to study the change in DOC and total (TOC) organic carbon concentrations in the main rivers of Switzerland (Rhône, Rhine, Thur and Aar) between 1974 and 2010. These rivers are characterized by high discharge regimes (due to their Alpine origin) and by running in populated areas. Small long term trends (a general statistically significant decrease in TOC and a less clear increase in DOC concentrations), on the order of 1% of mean OC concentration per year, have been observed. An upward trend before 1999 reversed direction to a more marked downward trend from 1999 to 2010. Of the potential causes of OC temporal variation analysed (water temperature, dissolved reactive phosphorus and river discharge), only discharge explains a significant, albeit still small, part of TOC variability (8-31%), while accounting for barely 2.5% of DOC variability. Estimated anthropogenic TOC and DOC loads (treated sewage) to the rivers could account for a maximum of 4-20% of the temporal trends. Such low predictability is a good example of the limitations faced when studying causality and drivers behind small variations in complex systems. River export of OC from Switzerland has decreased significantly over the period. Since about 5.5% of estimated NEP of Switzerland is exported by the rivers, riverine OC fluxes should be taken into account in a detailed carbon budget of the country.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphor, rot, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, puriss. p.a., powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphor, rot, ≥97.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Aktivkohle, suitable for GC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.