Merck
  • Spatial and temporal variability of organic C and N concentrations and export from 30 boreal rivers induced by land use and climate.
Spatial and temporal variability of organic C and N concentrations and export from 30 boreal rivers induced by land use and climate.
The Science of the total environment (2015-01-06)
Tuija Mattsson, Pirkko Kortelainen, Antti Räike, Ahti Lepistö, David N Thomas
ZUSAMMENFASSUNG
Climate change scenarios for northern boreal regions indicate that there will be increasing temperature and precipitation, and the changes are expected to be larger in winter than in summer. These precipitation and discharge patterns, coupled with shorter ice cover/soil frost periods in the future would be expected to contribute significantly to changing flow paths of organic matter over a range of land use patterns. In order to study the impact of climate change on the seasonality of organic matter export we compared total organic carbon (TOC) and total organic nitrogen (TON) concentrations and export, during different seasons and climatically different years, over 12 years for 30 Finnish rivers separated into forest, agriculture and peat dominated catchments. The mean monthly TOC concentrations were highest during autumn and there was also a peak in May during the highest flow period. The mean monthly concentrations of TON were lowest during winter, increased in spring and remaining high throughout summer and autumn. The TOC/TON ratios were lowest during summer and highest during winter, and in all seasons the ratios were lowest in catchments with a high proportion of agricultural land and highest in peat-dominated catchments. The seasonality of TOC and TON exports reflected geographical location, hydrology and land use patterns. Most of the TOC and TON were transported during the high flow following the spring snowmelt and during rainfall in autumn. In all catchments the relative importance of the spring snowmelt decreased in wet and warm years. However, in peat-dominated catchments the proportion of spring period was over 30% of the annual export even in these wet and warm years, while in other catchments the proportion was about 20%. This might be linked to the northern location of the peat-dominated catchments and the permanent snow cover and spring snowmelt, even in warm years.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, puriss. p.a., powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Stickstoff, ≥99.998%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Aktivkohle, suitable for GC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Kohlenstoff, foil, 5x5mm, thickness 2.0mm, hOpg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.