Merck
  • Synthesis, characterization, and in vitro studies of PLGA-PEG nanoparticles for oral insulin delivery.
Synthesis, characterization, and in vitro studies of PLGA-PEG nanoparticles for oral insulin delivery.
Chemical biology & drug design (2014-04-02)
Sara Hosseininasab, Roghiyeh Pashaei-Asl, Amir Ahmad Khandaghi, Hamid Tayefi Nasrabadi, Kazem Nejati-Koshki, Abolfazl Akbarzadeh, Sang Woo Joo, Younes Hanifehpour, Soodabeh Davaran
ZUSAMMENFASSUNG
Therapeutic proteins and peptides are corresponding to a major area of research in biotechnology companies and current pharmaceutical. Because of their natural instability, the enormous majority of these drugs require parentéral administration. Oral insulin delivery would be a highly attractive alternative process of administration, though it continues to be a mysterious target due to the enzymatic digestion of insulin and low levels of absorption from the gastrointestinal region. Hydrogel polymers can be considered as potential carriers for oral insulin delivery. In particular, a pH responsive hydrogel composed of PLGA-PEG has shown the ability to protect insulin from enzymes in the gastric environment and release in small intestines. However, this material has not shown similar potential for oral protein delivery of further model drugs. To date, the unique interaction between PLGA-PEG and insulin, as a potential drug for oral delivery, is not completely understood. The focus of this research is synthetization and characterization of hydrogels PLGA-PEG insulin nanoparticles and also pH sensitivity of insulin nanoparticles was investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Zinn(II)-2-Ethylhexanoat, 92.5-100.0%
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Dichlormethan, Selectophore, ≥99.5%
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material