Merck
  • Preparation, characterization, and antitumor activities of folate-decorated docetaxel-loaded human serum albumin nanoparticles.
Preparation, characterization, and antitumor activities of folate-decorated docetaxel-loaded human serum albumin nanoparticles.
Drug delivery (2014-01-30)
Shougang Jiang, Xianfeng Gong, Xiuhua Zhao, Yuangang Zu
ZUSAMMENFASSUNG
Docetaxel is now a major antitumor drug in clinical use for the treatment of a variety of tumors. The ethanol/Tween 80 solvent required in the formulation to increase the docetaxel solubility is at least partly responsible for the hypersensitivity reaction, decreased uptake by tumor tissue, and increased exposure to other body compartments. The present study was aimed at developing hydrosoluble DTX-FA-HSANPs targeting tumor cells and to investigate antitumor activities of the nanoparticles. The DTX-HSANPs were prepared using a desolvation technique and the carboxylic groups of NHS-folate were conjugated with the amino groups of the human serum albumin nanoparticles, and studied their size and zeta potential, drug loading efficiency, surface morphology, release properties in vitro, and antitumor activities. The spherical nanoparticles obtained were negatively charged with a zeta potential of about -30 mV and characterized around 150 nm with a narrow size distribution. Drug loading efficiency was approximately 17.2%. The folate-decorated nanoparticles targeted a human hepatoma cell line effectively. The in vitro drug release of DTX-FA-HSANPs in the first 96 h corresponded with the following equation: Q = 18.87851 - 0.13866t + 0.21276t² - 0.00704t³ + 0.0000847854t⁴ - 0.00000034991t⁵ (R² = 0.98155). Moreover, the in vitro antitumor activities of DTX-FA-HSANPs were close to the activities of the positive control (docetaxel). The in vivo inhibition ratios of DTX-FA-HSANPs and docetaxel were 66.2% and 59.5%, respectively, at a dose of 5 mg/kg. In light of the observed antitumor activities, it would be of considerable interest to collect sufficient data for the clinical application of docetaxel-loaded nanoparticles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard